Pyronin G
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 92-32-0 Molecular Formula: C17H19ClN2O Formula Weight: 302.80 Physical Form: Crystalline powder Appearance: Green Melting Point: 250-260° Beilstein Reference: 3795789 Merck Reference: 14,8007 MDL No.: MFCD00011725
Marker dye for RNA useful in acid buffer systems
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS Number:92-32-0
- Formula:C₁₇H₁₉ClN₂O
- MDL Number:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:14,8007
- Melting Pt:250…260 °C
- Storage Temperature:Ambient
- EINECS:202-147-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



