Pyronin G
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 92-32-0 Molecular Formula: C17H19ClN2O Formula Weight: 302.80 Physical Form: Crystalline powder Appearance: Green Melting Point: 250-260° Beilstein Reference: 3795789 Merck Reference: 14,8007 MDL No.: MFCD00011725
Marker dye for RNA useful in acid buffer systems
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS Number:92-32-0
- Formula:C₁₇H₁₉ClN₂O
- MDL Number:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:14,8007
- Melting Pt:250…260 °C
- Storage Temperature:Ambient
- EINECS:202-147-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



