Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyronin G

Pyronin G

Glass bottle for solids, 1 g
Catalog # J61068.03
  
Pyronin G
Catalog # J61068.03
  
CAS Number:  92-32-0
Pyronin G
Pyronin G

Glass bottle for solids, 1 g

CAS Number:  92-32-0
Price
35,90 €
Log in for availability
1 * 1 g
Product Details & Documents

CAS No.: 92-32-0 Molecular Formula: C17H19ClN2O Formula Weight: 302.80 Physical Form: Crystalline powder Appearance: Green Melting Point: 250-260° Beilstein Reference: 3795789 Merck Reference: 14,8007 MDL No.: MFCD00011725

Marker dye for RNA useful in acid buffer systems

Synonyms:  Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
  • CAS Number:92-32-0
  • Formula:C₁₇H₁₉ClN₂O
  • MDL Number:MFCD00011725
  • Molecular Weight:302.8 g/mol
  • Merck Index:14,8007
  • Melting Pt:250…260 °C
  • Storage Temperature:Ambient
  • EINECS:202-147-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Cat. No.:
    J61068.03

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.