Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 2 L
Supplier: Thermo Fisher Scientific
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # J62449.K3
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236358
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2 L
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
- Cat. No.:J62449.K3
- Physical form:Liquid
- MDL No.:MFCD00236358
- Appearance:Colourless



