Buffer, potassium phosphate, 0,2 M solution
Potassium phosphate, 0,2M buffer solution, pH 8,0, Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Buffer, potassium phosphate, 0,2 M solution
Catalog # J60851.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:Potassium phosphate, 0,2M buffer solution, pH 8,0
- Cat. No.:J60851.AP
- Physical form:Liquid
- MDL No.:MFCD00011401
- CAS No.:7778-77-0



