Mesenchymal stem cell differentiation medium
Mesenchymal Stem Cell Chondrogenic Differentiation Medium (ready-to-use), 100 ml, 100 mL
Catalog # C-28012
Marketsource
Supplier: PROMOCELL
Mesenchymal stem cell differentiation medium
Catalog # C-28012
Supplier: PROMOCELL
Marketsource
CAS Number:
Jump to:
Product Details & Documents
PromoCell Mesenchymal Stem Cell Differentiation Media efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively.
Specifications
Specifications
- Pk:100 mL
- Description:Mesenchymal Stem Cell Chondrogenic Differentiation Medium (ready-to-use), 100 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



