Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 1 L
Catalog # J61544.K2
Tween 20 blocking buffer, 1% in PBS (10X)
Catalog # J61544.K2
CAS Number:  
Tween 20 blocking buffer, 1% in PBS (10X)
Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 1 L

CAS Number:  
Price
86,60 €
Log in for availability
1 * 1 L
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid
Appearance: Clear

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Tween 20 Blocking Buffer, 1% in PBS (10X)
  • Cat. No.:
    J61544.K2
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Appearance:
    Clear

Specifications