Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 500 mL
Catalog # J61544.AP
Tween 20 blocking buffer, 1% in PBS (10X)
Catalog # J61544.AP
CAS Number:  
Tween 20 blocking buffer, 1% in PBS (10X)
Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 500 mL

CAS Number:  
Price
65,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid
Appearance: Clear

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Tween 20 Blocking Buffer, 1% in PBS (10X)
  • Cat. No.:
    J61544.AP
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Appearance:
    Clear

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.