Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, potassium phosphate, 0,2 M solution

Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,5, Glass bottle, 1 L
Catalog # J62239.K2
Buffer, potassium phosphate, 0,2 M solution
Catalog # J62239.K2
CAS Number:  
Buffer, potassium phosphate, 0,2 M solution
Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,5, Glass bottle, 1 L

Wechseln zu:
CAS Number:  
Price
122,00 €
Anmelden für die Verfügbarkeit
1 * 1 L
Product Details & Documents

CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401

Spezifikationen:

Spezifikationen:

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Potassium phosphate, 0,2M buffer solution, pH 7,5
  • Cat. No.:
    J62239.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00011401
  • CAS No.:
    7778-77-0

Spezifikationen:

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.