Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DB-10C DRY BLOCK TERMOSTAT FOR 10 X STANDARD 10MMX10MM CUVETTES 25-42C, TIMER 1M TO 96 H 1 * 1 items
Catalog # 460-0194
Supplier:
GRANT INSTRUMENTS
Expand image
DB-10C DRY BLOCK TERMOSTAT FOR 10 X STANDARD 10MMX10MM CUVETTES 25-42C, TIMER 1M TO 96 H 1 * 1 items
Catalog # 460-0194
Supplier:
GRANT INSTRUMENTS