Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]QUALITATIVE FLAT FILTER 450MM 6-10µm 1 * 100 ST
Catalog # PRATA045110
Supplier:
PRAT DUMAS
Expand image
[EN]QUALITATIVE FLAT FILTER 450MM 6-10µm 1 * 100 ST
Catalog # PRATA045110
Supplier:
PRAT DUMAS