ContactUsLinkComponent
Plug screwed, bore size 5.7mm, polypropylene, for use with Torion glass bodies and rubber/PTFE sealing rings, fittings available in choice of materials, glass filled PP for use at temperatures up to 180C. 1 * 10 items
Artikelnr. BIBB720-01
Supplier:  SciLabware
Plug screwed, bore size 5.7mm, polypropylene, for use with Torion glass bodies and rubber/PTFE sealing rings, fittings available in choice of materials, glass filled PP for use at temperatures up to 180C. 1 * 10 items
Artikelnr. BIBB720-01
Supplier:  SciLabware
Frequently Bought Together