ContactUsLinkComponent
Sucrose (Brix) Standard 10% - 1.347824 (± 0.00004), Stabilised, shelf-life of 1 year, 6 x 15ml 1 * 90 mL
Catalog # REGEBS10S6
Supplier:  REAGECON
Sucrose (Brix) Standard 10% - 1.347824 (± 0.00004), Stabilised, shelf-life of 1 year, 6 x 15ml 1 * 90 mL
Catalog # REGEBS10S6
Supplier:  REAGECON
Frequently Bought Together