Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sucrose (Brix) Standard 11.2% - 1.349682 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS112S
Supplier:
REAGECON
Expand image
Sucrose (Brix) Standard 11.2% - 1.349682 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS112S
Supplier:
REAGECON
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals