Safety spectacles, XC™
Anti-scratch, Blue, Dark TCG PC
DALL1012910
The large curved lens of these safety glasses offers a total wrap-around coverage and provides exceptional peripheral vision along with complete protection.
- Adjustable arm length and lens angle
- Pads for excellent non slip function
- Extra flexible frame; for men and women
- Multi-materials provide extra comfort; soft brow, nose bridge and temples
EN 166, EN 170
: EN 166, EN 170
- Type:Anti-scratch
- Colour:Blue
- Seq:11
- Lenses:Dark TCG PC