Orders LinkContactUsLinkComponent
Safety spectacles, XC™
Safety spectacles, XC™
Anti-scratch, Blue, HDL yellow PC
Catalog # DALL1011024
Supplier:   HONEYWELL SAFETY
Safety spectacles, XC™
Catalog # DALL1011024
Supplier:  HONEYWELL SAFETY
Safety spectacles, XC™
Safety spectacles, XC™
Anti-scratch, Blue, HDL yellow PC
Price
22,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The large curved lens of these safety glasses offers a total wrap-around coverage and provides exceptional peripheral vision along with complete protection.
  • Adjustable arm length and lens angle
  • Pads for excellent non slip function
  • Extra flexible frame; for men and women
  • Multi-materials provide extra comfort; soft brow, nose bridge and temples
EN 166, EN 170
Certifications: EN 166, EN 170
Specifications
  • Type:
    Anti-scratch
  • Colour:
    Blue
  • Seq:
    05
  • Lenses:
    HDL yellow PC
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Adapter corrective lenses for safety spectacles, XC™
    Adapter corrective lenses for safety spectacles, XC™
    111-3720
    Log in for availability
    1 * 1 ST - 50,70 €