Orders LinkContactUsLinkComponent
Safety spectacles, XC™
Anti-scratch, Blue, I/O silver PC
  DALL1011023
Safety spectacles, XC™
  DALL1011023
Safety spectacles, XC™
Safety spectacles, XC™
Anti-scratch, Blue, I/O silver PC
25,50 €
1 * 1 ST
The large curved lens of these safety glasses offers a total wrap-around coverage and provides exceptional peripheral vision along with complete protection.
  • Adjustable arm length and lens angle
  • Pads for excellent non slip function
  • Extra flexible frame; for men and women
  • Multi-materials provide extra comfort; soft brow, nose bridge and temples
EN 166, EN 170
Certifications: EN 166, EN 170
  • Type:
    Anti-scratch
  • Colour:
    Blue
  • Seq:
    04
  • Lenses:
    I/O silver PC
Accessories
  •  
    Description
  • 111-3720
    Adapter corrective lenses for safety spectacles, XC™
    1 * 1 ST - 50,70 €