Orders LinkContactUsLinkComponent
Trypsin neutralizing solution
Trypsin neutralising solution (TNS), 125 mL
  C-41110
Marketsource
 :   PROMOCELL
Trypsin neutralizing solution
  C-41110
 :  PROMOCELL
Marketsource
 :  
Trypsin neutralizing solution
Trypsin neutralizing solution
Trypsin neutralising solution (TNS), 125 mL
 :  
114,00 €
1 * 125 mL
Trypsin is the most widely employed enzyme used for cell detachment. This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0,05% trypsin inhibitor in 0,1% BSA.
 : For in vitro research use only and not for diagnostic or therapeutic procedures.
  • Pk:
    125 mL
  • Description:
    Trypsin neutralising solution (TNS)