Orders LinkContactUsLinkComponent
Accessories
Accessories
Port, Ø 14 mm, can be closed by flap, custom positions in the back wall, IN75plus
Catalog # MEMMIN75PLUSD6
Supplier:   MEMMERT
Accessories
Catalog # MEMMIN75PLUSD6
Supplier:  MEMMERT
Accessories
Port, Ø 14 mm, can be closed by flap, custom positions in the back wall, IN75plus
Price
220,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Port, Ø 14 mm, can be closed by flap, custom positions in the back wall
  • Seq:
    16
  • For:
    IN75plus