Sample containers, straight, with screw cap, WHEATON®
150 ml, No label, sterile, Clear, PS/metal
Supplier: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Catalog # 216-1015
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Specifications
- Type:No label, sterile
- Capacity:150 ml
- Colour:Clear
- Material:PS/metal