Orders LinkContactUsLinkComponent
Counting chambers, Neubauer
Counting chambers, Neubauer
Counting chamber, bright-lined, Neubauer, 0,10 mm
Catalog # MARI0640130
Supplier:   MARIENFELD
Counting chambers, Neubauer
Catalog # MARI0640130
Supplier:  MARIENFELD
Counting chambers, Neubauer
Counting chamber, bright-lined, Neubauer, 0,10 mm
Price
73,76 €
Log in for availability
1 * 1 ST
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
Certifications: IVD 98/79/EC
Delivery information: Supplied with two cover glasses.
Documents
Specifications
  • Depth of chamber:
    0,10 mm
  • Description:
    Counting chamber, bright-lined, Neubauer
  • Seq:
    02