Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
7 COMPOSES 1000 MG/L CHAQUE Lithium (Li+) Sodium (Na+) Ammonium (NH4+) Potassium (K+) Magnesium (Mg2+) Calcium (Ca2+) Strontium (Sr2+) DANS H2O 1 * 100 ml
Catalog # 89254.180
Supplier:
VWR CUSTOM MADE CHEMICALS
Bild vergrößern
7 COMPOSES 1000 MG/L CHAQUE Lithium (Li+) Sodium (Na+) Ammonium (NH4+) Potassium (K+) Magnesium (Mg2+) Calcium (Ca2+) Strontium (Sr2+) DANS H2O 1 * 100 ml
Catalog # 89254.180
Supplier:
VWR CUSTOM MADE CHEMICALS