Rectangular cover glasses, Corning®
22×30 mm, #1,5
Supplier: Corning
Rectangular cover glasses, Corning®
Catalog # CORN2980-223
Supplier: Corning
Jump to:
Product Details & Documents
These rectangular cover glasses are made from borosilicate glass.
Documents
Certificates
Search by Lot NumberSpecifications
- No.:#1,5
- L×W:22×30 mm
- Seq:01
- Cat. No.:CORN2980-223
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact