Orders LinkContactUsLinkComponent
Rectangular cover glasses, Corning®
Rectangular cover glasses, Corning®
22×50 mm, #1,5
Catalog # CORN2980-225
   Low Manufacturing Impact
Supplier:   Corning
Rectangular cover glasses, Corning®
Catalog # CORN2980-225
Supplier:  Corning
   Low Manufacturing Impact
Rectangular cover glasses, Corning®
Rectangular cover glasses, Corning®
22×50 mm, #1,5
Price
306,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These rectangular cover glasses are made from borosilicate glass.
Documents
Specifications
  • No.:
    #1,5
  • L×W:
    22×50 mm
  • Seq:
    03
  • Cat. No.:
    CORN2980-225
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact