n-Eicosane ≥99%
Glass bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 112-95-8 Molecular Formula: CH3(CH2)18CH3 Formula Weight: 282.54 Flash Point: 176°(348°F) Refractive Index: 1.4425 Melting Point: 36-38° Boiling Point: 220°/30mm Beilstein Reference: 1700722 MDL No.: MFCD00009344
Synonyms: N-Eicosane
- CAS Number:112-95-8
- Formula:H₃C(CH₂)₁₈CH₃
- MDL Number:MFCD00009344
- Molecular Weight:282.55 g/mol
- Boiling Pt:220 °C (30 mmHg)
- Melting Pt:36…38 °C
- Density:0.7888
- Flash Pt:176 °C (348 °F)
- Storage Temperature:Ambient
- EINECS:204-018-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



