Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TWEEZER ROUND TIP STYLE 2ADE 10X1.5X35MM 1 * 1 items
Catalog # OUTI2ADE
Supplier:
OUTILS RUBIS
Expand image
TWEEZER ROUND TIP STYLE 2ADE 10X1.5X35MM 1 * 1 items
Catalog # OUTI2ADE
Supplier:
OUTILS RUBIS