Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(4-Fluorophenoxy)benzaldehyde ≥97%

2-(4-Fluorophenoxy)benzaldehyde ≥97%

Glass bottle for solids, 1 g
Catalog # H34259.03
Toxic
Irritant
2-(4-Fluorophenoxy)benzaldehyde ≥97%
Catalog # H34259.03
CAS Number:  320423-61-8
2-(4-Fluorophenoxy)benzaldehyde ≥97%
2-(4-Fluorophenoxy)benzaldehyde ≥97%

Glass bottle for solids, 1 g

CAS Number:  320423-61-8
Price
78,30 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 320423-61-8 Molecular Formula: C13H9FO2 Formula Weight: 216.21 MDL No.: MFCD01568770

  • CAS Number:320423-61-8
  • Formula:C₁₃H₉FO₂
  • MDL Number:MFCD01568770
  • Molecular Weight:216.21 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications