Inlet liners
Liner, GC, ultra inert, straight, recommended for SPME (solid phase microextraction) injections, 0.75 mm
HEWL5190-4048
: Agilent
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
- Ø int.:0.75 mm
- Description:Liner, GC, ultra inert, straight, recommended for SPME (solid phase microextraction) injections
- Seq:02