Order Entry
Germany
Orders LinkContactUsLinkComponent
Quinoline ≥97%

Quinoline ≥97%

Glass bottle for solids, 200 kg
Catalog # A11545.PG
Toxic
Health
Irritant
Quinoline ≥97%
Catalog # A11545.PG
CAS Number:  91-22-5
Quinoline ≥97%
Quinoline ≥97%

Glass bottle for solids, 200 kg

CAS Number:  91-22-5
Price
Inquire For Price
Log in for availability
1 * 200 kg
Product Details & Documents

Purity: 97% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Refractive Index: 1.6260 Melting Point: -15° Boiling Point: 237° Solubility: More soluble in hot than cold water. Miscible with alcohol, ether Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736

  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:14,8068
  • Boiling Pt:237 °C
  • Melting Pt:–15 °C
  • Density:1.090
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 kg

Specifications