ContactUsLinkComponent
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 1-10 ?l/min for gradient 1 (Ka=5 Kb=3), 50-500 nl/min for gradient 2, and loading p ump for 1-30 ?l/min. Max. pressure 10,000 psi. Temperature controlled column compartment with two 10 port column swit 1 * 1 items
Catalog # 903-0608
Supplier:  EKSIGENT
NanoLC-Ultra 2Dplus system. Dual Gradient pump, 1-10 ?l/min for gradient 1 (Ka=5 Kb=3), 50-500 nl/min for gradient 2, and loading p ump for 1-30 ?l/min. Max. pressure 10,000 psi. Temperature controlled column compartment with two 10 port column swit 1 * 1 items
Catalog # 903-0608
Supplier:  EKSIGENT