ContactUsLinkComponent
NanoLC-Ultra 2D system. Dual Gradient pump: 50-500 nl/min for both gradients. Max. pressure 10,000 psi. Temperature controlled colum n compartment with two 10 port column switching valves. Includes NanoLC start-up kit (p/n 801-00043) and Eksigent con 1 * 1 items
Catalog # 903-0611
Supplier:  EKSIGENT
NanoLC-Ultra 2D system. Dual Gradient pump: 50-500 nl/min for both gradients. Max. pressure 10,000 psi. Temperature controlled colum n compartment with two 10 port column switching valves. Includes NanoLC start-up kit (p/n 801-00043) and Eksigent con 1 * 1 items
Catalog # 903-0611
Supplier:  EKSIGENT