Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer pack PCR Phire reaction final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml of 50mM MgCl 2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-524L
Supplier:
FINNZYMES REAGENTS
Expand image
Buffer pack PCR Phire reaction final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml of 50mM MgCl 2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-524L
Supplier:
FINNZYMES REAGENTS