Orders LinkContactUsLinkComponent
Test panels
Test panels
Test panel, cold rolled steel, smooth, degreased and cleaned, 60×200×0,5 mm
Catalog # SIMEVF8510
Supplier:   Industrial Physics
Test panels
Catalog # SIMEVF8510
Supplier:  Industrial Physics
Test panels
Test panels
Test panel, cold rolled steel, smooth, degreased and cleaned, 60×200×0,5 mm
Price
303,00 €
Log in for availability
1 * 400 ST
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
  • Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
  • Physical testing: Adhesion, elongation, impact resistance and elasticity
  • Appearance testing of coatings (gloss, colour, texture)
Documents
Specifications
  • Description:
    Test panel, cold rolled steel, smooth, degreased and cleaned
  • Seq:
    16
  • W×D×H:
    60×200×0,5 mm