Test panels
Test panel, cold rolled steel, smooth, degreased and cleaned, 75×150×0,5 mm
Catalog # SIMEVF8511
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Videos
Specifications
- Description:Test panel, cold rolled steel, smooth, degreased and cleaned
- Seq:17
- W×D×H:75×150×0,5 mm