Order Entry
Germany
Orders LinkContactUsLinkComponent
Capillaries for LC systems

Capillaries for LC systems

Capillary, stainless steel, 0,17×90 mm
Catalog # HEWL5067-4748
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWL5067-4748
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems

Capillary, stainless steel, 0,17×90 mm

Price
88,00 €
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.

Pre-swaged fittings are assembled according to Swagelok specifications.

Specifications
  • Description:
    Capillary, stainless steel, 0,17×90 mm
  • Seq:
    83
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.