Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Connection capillaries, stainless steel, 220 mm, I.Ø 0,12 mm, to connect TCC to DAD
Catalog # HEWL5067-4660
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWL5067-4660
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Connection capillaries, stainless steel, 220 mm, I.Ø 0,12 mm, to connect TCC to DAD
Price
150,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Connection capillaries, stainless steel, 220 mm, I.Ø 0,12 mm, to connect TCC to DAD
  • Seq:
    86