Solid phase extraction cartridges, Sep-Pak® Vac
Cartridge, C18, 37 - 55 µm, 3 ml, 200 mg
Catalog # WATEWAT054925
Marketsource
Supplier: WATERS
Solid phase extraction cartridges, Sep-Pak® Vac
Catalog # WATEWAT054925
Supplier: WATERS
Marketsource
Jump to:
Product Details & Documents
These cartridges deliver the option of minimum sample preparation. Consists of moulded, high purity PP syringe-barrel bodies with two PE frits to contain the packing material.
- Standard male Luer permits use with multi-position vacuum manifolds
- Colour coded printing on syringe barrels for identification
Specifications
- Column Format:Cartridge
- Particle Size:37 - 55 µm
- Capacity:3 ml
- Seq:04
- Bed weight:200 mg
- Phase:C18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



