Orders LinkContactUsLinkComponent
Stands for pipettes, Axygen®
Stands for pipettes, Axygen®
Carousel stand, holds 6 single channel pipettes (not compatible with Axygen Axypet Pro pipettors)
Catalog # AXYGAP-CSD
Supplier:   Corning
Stands for pipettes, Axygen®
Catalog # AXYGAP-CSD
Supplier:  Corning
Stands for pipettes, Axygen®
Stands for pipettes, Axygen®
Carousel stand, holds 6 single channel pipettes (not compatible with Axygen Axypet Pro pipettors)
Price
168,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Linear and carousel stands for one or several single or multi-channel pipettes.
Specifications
  • Description:
    Carousel stand, holds 6 single channel pipettes (not compatible with Axygen Axypet Pro pipettors)