ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, G6PDH (L. MESENTEROIDES), NAD ASSAYED, LYOPHILIZED. SAME AS CAT.# P5264 EXCEPT ASSAYED USING NAD. PHOSPH OHEXOSE ISOMERASE, PHOSPHOGLUCONATE DEHYDROGENASE, ADENYLATE KINASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES 1 * 2.000 EU
Catalog # ABNOP5267
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, G6PDH (L. MESENTEROIDES), NAD ASSAYED, LYOPHILIZED. SAME AS CAT.# P5264 EXCEPT ASSAYED USING NAD. PHOSPH OHEXOSE ISOMERASE, PHOSPHOGLUCONATE DEHYDROGENASE, ADENYLATE KINASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES 1 * 2.000 EU
Catalog # ABNOP5267
Supplier:  Abnova