ContactUsLinkComponent
KIT, PHOSPHATIDYL INOSITOL IGG/IGM ELISA KIT, PHOSPHATIDYL INOSITOL AB IGG/IGM ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QU ANTITATIVE MEASUREMENT OF IGG AND IGM CLASS AUTOANTIBODIES AGAINST PHOSPHATIDYL INOSITOL IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1092
Supplier:  Abnova
KIT, PHOSPHATIDYL INOSITOL IGG/IGM ELISA KIT, PHOSPHATIDYL INOSITOL AB IGG/IGM ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QU ANTITATIVE MEASUREMENT OF IGG AND IGM CLASS AUTOANTIBODIES AGAINST PHOSPHATIDYL INOSITOL IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1092
Supplier:  Abnova