Orders LinkContactUsLinkComponent
Personal eyewash
Personal eyewash
Strap for wall bracket, for extra support for the bottle when located in vibrating environments
Catalog # 118-0248
Supplier:   ORKLA CARE
Personal eyewash
Catalog # 118-0248
Supplier:  ORKLA CARE
Personal eyewash
Personal eyewash
Strap for wall bracket, for extra support for the bottle when located in vibrating environments
Price
12,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
  • Quick and easy to open
  • Eye cup protected from dust
  • Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
  • Neutralises the pH value
Expiry date: 4 years if container not opened.
* Bottle holder is supplied without eyewash.
Documents
Specifications
  • Description:
    Strap for wall bracket, for extra support for the bottle when located in vibrating environments
  • Seq:
    04