SPECIFICATIONS * OPTICS: All reflective for focusing at all wavelengths and precise imaging for micro samples * SOURCE: Ozone-free 15OW xenon lamp eliminates venting * SPECTROMETERS: Plane grating Czerny-Turner design maintains focus at all wavelengt 1 * 1 items
Catalog # JOBIFLUOROMAX-4
Supplier: JOBIN YVON EMISSION