Orders LinkContactUsLinkComponent
Incubator-clean
Incubator-clean
Incubator-Clean™ refill pack, Plastic container
Catalog # A5230.5000RF
Supplier:   PanReac AppliChem
Incubator-clean
Catalog # A5230.5000RF
Supplier:  PanReac AppliChem
Incubator-clean
Incubator-clean
Incubator-Clean™ refill pack, Plastic container
Price
545,46 €
Log in for availability
1 * 5 L
Product Details & Documents
Incubator-Clean™ disinfectant solution prevents contamination with, and growth of, fungi (and spores), bacteria (and spores, including Tuberculosis bacteria), viruses (including HIV and Hepatitis B) and mycoplasma in incubators and/or on sterile work benches. The active ingredients are quaternary benzylammonium compounds.
  • Free of mercury, formaldehyde, phenol or alcohol
  • Non corrosive to aluminium, tin-plated iron, chrome, nickel, steel, high grade steel and copper
  • Non toxic
Recommended use: Spray incubators once every 2 weeks; spray sterile benches once a day, or preferably before each laboratory worker begins to use the work area.
 : Use biocides safely. Always read the label and product information before use.
Specifications
  • Description:
    Incubator-Clean™ refill pack
  • Seq:
    03
  • Packed:
    Plastic container