Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Thioisonicotinamide

Glass bottle for solids, 1 g
Catalog # CAYM13056-1
Supplier:   Cayman Chemical
Thioisonicotinamide
Catalog # CAYM13056-1
Supplier:  Cayman Chemical
CAS Number:  2196-13-6
Thioisonicotinamide

Glass bottle for solids, 1 g

CAS Number:  2196-13-6
Price
18,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • MDL Number:MFCD00006437
  • Molecular Weight:138.19 g/mol
  • EINECS:218-592-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications