Order Entry
Germany
Orders LinkContactUsLinkComponent
O-Methylhydroxylamine hydrochloride

O-Methylhydroxylamine hydrochloride

Glass bottle for solids, 0,1 g
Catalog # CAYM400036-100
Supplier:   Cayman Chemical
O-Methylhydroxylamine hydrochloride
Catalog # CAYM400036-100
Supplier:  Cayman Chemical
CAS Number:  593-56-6
O-Methylhydroxylamine hydrochloride

Glass bottle for solids, 0,1 g

CAS Number:  593-56-6
Price
18,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
Synonyms:  Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
  • CAS Number:593-56-6
  • Formula:CH₆ClNO
  • MDL Number:MFCD00012951
  • Molecular Weight:83.515 g/mol
  • EINECS:209-798-7
  • UN:3261
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g

Specifications