Order Entry
Germany
Orders LinkContactUsLinkComponent
(S)-(+)-2-Hydroxyisovaleric acid

(S)-(+)-2-Hydroxyisovaleric acid

Glass bottle for solids, 500 mg
Catalog # CAYM10007090-500
Supplier:   Cayman Chemical
(S)-(+)-2-Hydroxyisovaleric acid
Catalog # CAYM10007090-500
Supplier:  Cayman Chemical
CAS Number:  17407-55-5
(S)-(+)-2-Hydroxyisovaleric acid

Glass bottle for solids, 500 mg

CAS Number:  17407-55-5
Price
46,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
  • CAS Number:17407-55-5
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066443
  • Molecular Weight:118.13 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.