Order Entry
Germany
Orders LinkContactUsLinkComponent
Estradiol

Estradiol

Glass bottle for solids, 1 g
Catalog # CAYM10006315-1
Supplier:   Cayman Chemical
Estradiol
Catalog # CAYM10006315-1
Supplier:  Cayman Chemical
CAS Number:  50-28-2
Estradiol

Glass bottle for solids, 1 g

CAS Number:  50-28-2
Price
68,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • MDL Number:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • EINECS:200-023-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.