Order Entry
Germany
Orders LinkContactUsLinkComponent
 

COD standards

COD standard solution, 1000 mg/l, 500 mL
Catalog # REGECOD1000
Supplier:   REAGECON
COD standards
Catalog # REGECOD1000
Supplier:  REAGECON
CAS Number:  
COD standards

COD standard solution, 1000 mg/l, 500 mL

CAS Number:  
Price
177,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Ideal for use as control standards to verify that correct analysis for COD has taken place.

Documents

Specifications

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    500 mL
  • Description:
    COD standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.