Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium nitrite ≥98%

Sodium nitrite ≥98%

Glass bottle for solids, 1.000 g
Catalog # A18668.0B
Toxic
Oxidizing
Environment
Sodium nitrite ≥98%
Catalog # A18668.0B
CAS Number:  7632-00-0
Sodium nitrite ≥98%
Sodium nitrite ≥98%

Glass bottle for solids, 1.000 g

CAS Number:  7632-00-0
Price
40,90 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118

Synonyms:  Diazotizing salts
  • CAS Number:7632-00-0
  • Formula:NaNO₂
  • MDL Number:MFCD00011118
  • Molecular Weight:69 g/mol
  • Merck Index:14,8648
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:271 °C (dec.)
  • Density:2.17
  • Storage Temperature:Ambient
  • EINECS:231-555-9
  • UN:1500
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.