Order Entry
Germany
Orders LinkContactUsLinkComponent
 

6-Hydroxyindole ≥98%

Glass bottle for solids, 5 g
Catalog # H55951.06
Corrosive
Irritant
Environment
6-Hydroxyindole ≥98%
Catalog # H55951.06
CAS Number:  2380-86-1
6-Hydroxyindole ≥98%
6-Hydroxyindole ≥98%

Glass bottle for solids, 5 g

CAS Number:  2380-86-1
Price
542,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
  • CAS Number:2380-86-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152101
  • Molecular Weight:133.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Ambient
  • EINECS:417-020-4
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications